VWR® WASHBLOCKER® Dike, Spill containment barriers
Supplier: VWR Collection
10147-836
VWRU10147-836EA 1800 SGD
VWRU10147-836
VWR® WASHBLOCKER® Dike, Spill containment barriers
Spill Control Materials
Flexible, low profile, portable 100% polyurethane barrier for containment of spillages on floors. Ideal for routine containment of washdown and sanitising liquid. Block doorways, protect carpeting, and channel liquid to drains.These reusable barriers are easily cleaned with soap and water.
  • Lightweight strip - easy to handle and store
  • Bright green colour is highly visible
  • Tear-resistant PP top withstands repeated use
  • Tacky urethane construction creates a leakproof seal on dry floors
Order Now
SPECIFICATIONS
Environmentally Preferable