VWR® A-Series, Analytical balances
Supplier: VWR Collection
10204-966
VWRU10204-966EA 5090 SGD
VWRU10204-966
VWR® A-Series, Analytical balances
Balances and Scales
A-series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
  • Internal calibration
  • Multiple weigh modes
  • USB (A and B) communication interfaces and 2×RS232
  • Easy-to-read, adjustable, backlit display
These economical balances offer a variety of standard features, such as internal calibration, GLP procedures, 17 weighing units, adjustable ambient condition settings, 11 application modes, and multilingual operation. The VWR® A-Series balances are the answer for organisations demanding precision andMore Product Information