VWR®, Beakers, Low Form, Heavy Duty
Supplier: VWR Collection
10536-390 10536-404 10536-402 10536-392 10536-518
VWRU10536-390EA 249 SGD
VWRU10536-390 VWRU10536-404 VWRU10536-402 VWRU10536-392 VWRU10536-518
VWR®, Beakers, Low Form, Heavy Duty
Beakers
VWR® low form, heavy duty beakers are mechanically stronger than standard beakers and are designed for longer life with increased user safety.
  • With reinforced rim and spout
  • Borosilicate glass fabrication
  • Thick wall and heavy base
Durable matte finish marking area and graduated scale printed in white colour.
Certifications: Manufactured in accordance to ASTM E960 Type-2 specification.