VWR®, Büchner Funnels
Supplier: VWR Collection
89000-444 10545-896
VWRU89000-444EA 196 SGD
VWRU89000-444 VWRU10545-896
VWR®, Büchner Funnels
Funnels Filtration Funnels Büchner Funnels
Borosilicate glass 3.3.
  • With KIMFLOW® fritted disc
  • With heavy walled stems
  • With marking spot
  • Beaded rims for extra strength
This funnel is used wherever the filter paper and membranes are unable to withstand the chemical attack of the filtrate.