VWR®, Graduated Pipettes, Class A
Supplier: VWR Collection
10546-014 10546-024 10546-006 10546-008 10546-036 10546-028 10546-032 10546-010 10546-012 10546-026 10546-018
VWRU10546-014EA 106 SGD
VWRU10546-014 VWRU10546-024 VWRU10546-006 VWRU10546-008 VWRU10546-036 VWRU10546-028 VWRU10546-032 VWRU10546-010 VWRU10546-012 VWRU10546-026 VWRU10546-018
VWR®, Graduated Pipettes, Class A
Pipettes Graduated Pipettes
Borosilicate 3.3 expansion glass, class A.
  • Reusable
  • Colour coded for easy sorting
Calibrated at 20 °C to deliver. These pipettes are unserialised or batch certified.
Certifications: Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969. Pipettes are colour coded as per ASTM specification E1273.