VWR®, Powder Funnels, Glass
Supplier: VWR Collection
10545-910 10546-046 10546-048 10545-912
VWRU10545-910EA 178 SGD
VWRU10545-910 VWRU10546-046 VWRU10546-048 VWRU10545-912
VWR®, Powder Funnels, Glass
Funnels Powder Funnels
Made from Type 1 Class A borosilicate 3.3 expansion glass.
  • Constructed with a heavy, uniform wall
  • Fire-polished rim and stem to provide long service life
  • Chemical resistant
  • White printing
Funnels comes with short, wide stem, and are useful for adding powdered chemical to reaction vessels.
Certifications: In acc. with ASTM E-1095 standards.
Order Now
SPECIFICATIONS
Environmentally Preferable