VWR®, Volumetric Flasks, Class A, with Glass Stopper
Supplier: VWR Collection
89000-414 89000-404 89000-400 89000-402 89000-412 10546-066
VWRU89000-414EA 131 SGD
VWRU89000-414 VWRU89000-404 VWRU89000-400 VWRU89000-402 VWRU89000-412 VWRU10546-066
VWR®, Volumetric Flasks, Class A, with Glass Stopper
Flasks Volumetric Flasks
Manufactured from 3.3 expansion, low extractable borosilicate glass with standard taper glass stopper.
  • Calibrated to contain
  • With pennyhead glass stopper
  • Flasks feature a single graduation ring blasted on the neck and a marking spot
Sizes 5 ml and larger are designed from ASTM Specification E288, Class A requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.