VWR® Easyplate™, Petri dish racks
Supplier: VWR Collection
10600-106
VWRU10600-106EA 2040 SGD
VWRU10600-106
VWR® Easyplate™, Petri dish racks
Racks Petri Dish Racks
Made of lightweight, chemical resistant polycarbonate. The four-channel design accommodates stacks of 100 mm diameter Petri dishes. They have silicone edges on the channels which allow random entry and retrieval of dishes, and hold them securely in place.
  • Center channel maximises CO₂ flow through rack
  • Increases sample capacities of incubators and benchtops
  • Simplifies the organisation, storage, and transport of Petri dishes
Ordering information: Each assorted colours pack includes one rack in each of white, red, yellow, and blue.