VWR®, Disposable Goggles, Sterile
Supplier: VWR Collection
10770-152
VWRU10770-152EA 1010 SGD
VWRU10770-152
VWR®, Disposable Goggles, Sterile
Goggles
Sterile disposable goggles are individually bagged and provide optimum comfort for extended wear times.
  • Two types of ventilation
  • Anti-fog and anti-scratch
  • Fits over prescription glasses
  • PVC frame
  • Polycarbonate lens
These goggles come in two types of ventilation: indirect and direct. Produced under stringent conditions and monitored closely to ensure high levels of quality for every pair. The goggles passed tests according to the American National Standard for Occupational and Educational Personal Eye and Face Protective devices. Ideal for use inMore Product Information