VWR®, Disposable, Anti-Static Weighing Boats
Supplier: VWR Collection
10770-452
VWRI611-9171EA 41.7 SGD
VWRI611-9171 VWRI611-9172 VWRI611-9173 VWRI611-9174 VWRU10770-452
VWR®, Disposable, Anti-Static Weighing Boats
Boats Weighing Boats
PS, hexagonal.
  • Flat bottom avoids tipping
  • Sloping sides provide accurate transfer
  • Temperature resistant up to 70 °C
These boats feature smooth surfaces to enable accurate pour-out with minimal sample loss. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases. These flexible boats bend at points to form pouring spouts.
Packaging: These weighing boats are produced and packaged in plastic bags in a cleanroom.
Order Now
SPECIFICATIONS
Environmentally Preferable