VWR®, Aprons, PE
Supplier: VWR Collection
10826-007
VWRU10826-007EA 219 SGD
VWRU10826-007
VWR®, Aprons, PE
Disposable Garments
This PE apron helps to keep clothing clean and dry.
  • Disposable
  • Medium-weight fabric
  • One-piece bib design just slips over the head and ties in back
Packaging: 100/dispenser box.
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application
Thickness 0,05 mm