VWR® Cleanroom Wipes, 100% Polyester
Supplier: VWR Collection
10835-982
VWRU10835-982EA 48.6 SGD
VWRU10835-982
VWR® Cleanroom Wipes, 100% Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These are 100% continuous-filament, double-knit polyester fabric wipes.
  • Free from silicon, amides, and DOP contamination
  • Extremely low particle and fiber levels
  • Laser cut edges
  • Absorbant
Resistant to abrasion, providing maximum cleaning performance. Designed for cleaning surfaces and controlling spills in ISO Class 4 cleanroom environments.