VWR®, Multiwell Cell Culture Plates
Supplier: VWR Collection
10861-564
VWRU10861-564EA 218 SGD
VWRU10861-564
VWR®, Multiwell Cell Culture Plates
Microplates Cell Culture Plates
Ideal for cell growth and cell yields.
  • Uniform well volume
  • Flat well bottom and round bottom plates available
  • Well surface is smooth and free from striation
  • Alphanumerically coded for easy identification
  • Sterilised by gamma irradiation, non-pyrogenic
Raised rims on wells with the uniform rings on the lid help to reduce evaporation.
Plates are designed for use in applications where cell attachment is not desired. Plates are not autoclavable.
Single-position lid reduces the risk of cross-contamination and handling mistakes.
Certifications: ISO13More Product Information