VWR®, Jars, Clear and Straight Sided
Supplier: VWR Collection
10861-934
VWRU10861-934EA 87.5 SGD
VWRU10861-934
VWR®, Jars, Clear and Straight Sided
Jars General Use Jars
VWR® straight sided jars are manufactured from soda-lime glass that conforms to USP Type III requirements.