VWR®, French Square Bottles, Clear
Supplier: VWR Collection
10862-150
VWRU10862-150EA 78.3 SGD
VWRU10862-150
VWR®, French Square Bottles, Clear
Bottles Glass Bottles
Made of soda-lime glass.
  • Optimised shelf space
  • Ideal for storage of solids
  • Wide-mouth design for efficient addition and removal of contents
The square shape maximises use of shelf space and facilitates storage. The clear design allows for constant viewing of bottle contents.
Certifications: Type III requirements.
Packaging: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 576 per wrap) packaged in corrugatedMore Product Information