VWR®, Wash Bottles, Narrow or Wide Neck, with Cap
Supplier: VWR Collection
16650-537 11215-302
VWRU16650-537EA 87.9 SGD
VWRU16650-537 VWRU11215-302
VWR®, Wash Bottles, Narrow or Wide Neck, with Cap
Bottles Wash Bottles
Bottles, LDPE red and caps, PP, red, are vividly coloured to aid in the identification of hazardous contents.
  • Good chemical resistance
  • Good temperature resistance