VWR®, Autoclavable Biohazard Bags
Supplier: VWR Collection
11215-826
VWRU11215-826EA 366 SGD
VWRU11215-826
VWR®, Autoclavable Biohazard Bags
Bags Waste Disposal Bags
These heavy-duty bags, in PE, with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic. Ideal for sterilising animal cages.
  • Clearly labeled with proper biohazard warnings
  • Bright red coloured for quick identification
  • Autoclavable up to 121 °C (250 °F)
Bags meet many state and local regulations.
Certifications: Bags meet many state and local regulations.
Delivery information: Supplied with 20,3 cm twist ties.