VWR® SECURE-T, Blender bags Stomacher® with tear-off top
Supplier: VWR Collection
11216-892
VWRU11216-892EA 503 SGD
VWRU11216-892
VWR® SECURE-T, Blender bags Stomacher® with tear-off top
Bags Blender Bags
Bags made of transparent polypropylene with wide-sealed bottom, designed to withstand the paddle action of stomacher lab blenders. Closure bags permit collection, transportation, blending, and storage of samples all within the same bag.
  • Heat extruded polyethylene tubing guarantees internal sterility
  • Maximum bag mouth opening to facilitate insertion of sample
  • Bags are malleable yet durable, with superior wall strength
  • Shrink-wrapped and features patented tear-off sterile bag
Order Now
SPECIFICATIONS
Environmentally Preferable