VWR®, Balance Tables
Supplier: VWR Collection
12567-330
VWRU12567-330EA 2920 SGD
VWRU12567-330
VWR®, Balance Tables
Furniture Tables and Workbenches
Constructed of four separate heavy-duty structural elements - two sturdy pedestal bases with non-skid rubber feet, a rigid cross-brace, and a chemical-resistant top. Chemical-resistant finish eliminate troublesome vibrations that affect the accuracy of analytical or microbalances with a balance table.
  • Rigid construction
  • Fast and easy assembly
  • Pedestal bases are hollow, so they can be filled with sand for even greater stability
Caution: Parts have to be assembled.