VWR®, Incubating Microplate Shaker
Supplier: VWR Collection
12620-932
VWRI444-0272EA 3720 SGD
VWRI444-0272 VWRI444-0273 VWRI444-7082 VWRU12620-932
VWR®, Incubating Microplate Shaker
Shakers Shaking Incubators
Incubating microplate shaker with microprocessor control which holds up to four microplates or two microtube racks. The speed can be adjusted across a wide range to suit the application; a low speed ensures that the well contents are not ejected, more vigorous agitation provides effective aeration across the small surface area of each of the wells. Ideal for use in immunoassays, hybridisations, biotechnology, microbiology and pharmacology applications.
  • Three independent LED displays show speed, temperature and time
  • Integral timer, 1 s to 160 h, shows
More Product Information