VWR® Dyla-Dual™, Hotplate stirrers
Supplier: VWR Collection
12620-976
VWRU12620-976EA 792 SGD
VWRU12620-976
VWR® Dyla-Dual™, Hotplate stirrers
Stirrers Magnetic Hotplate Stirrers
These economical and durable stirrers are ideal for general heating and stirring. A large cast aluminium top plate provides an even heating surface. Electronic speed control and a bimetallic thermostat deliver reliable and repeatable results. Powerful heater reaches maximum temperature in minutes.
Built-in support rod holder with locking knob accepts optional support rod and clamp kit for mounting thermometers and temperature probes. Large magnet provides strong magnetic coupling to the stirring bar. Indicator lights illuminate when unit is on.
Delivery information: Supplied with 457More Product Information