VWR® Traceable®, Refrigerator and Freezer Plus™ thermometers
Supplier: VWR Collection
12777-838
VWRU12777-838EA 217 SGD
VWRU12777-838
VWR® Traceable®, Refrigerator and Freezer Plus™ thermometers
Thermometers Digital Thermometers
Mercury-free, solid-state bottle probe in nontoxic glycol with three meter cable. These digital thermometers are designed for monitoring temperatures in freezers, water baths, heating blocks, incubators and refrigerators with this enclosed temperature-buffered sensor.
  • Monitors high and low readings overnight or for any time period
  • Programmable alarm when temperature rises above or falls below a set point
  • Triple display simultaneously shows MIN/MAX and current temperatures
Bottle insulates sensor from rapid temperature changes when refrigerator door isMore Product Information