VWR® Talon®, Support stand, tripod base
Supplier: VWR Collection
12985-076
VWRU12985-076EA 217 SGD
VWRU12985-076
VWR® Talon®, Support stand, tripod base
Retort Stands
Tripod based support stand for use with VWR® Talon® lab frames.
  • Support stand base is constructed of cast iron with black enamel finish
  • Base features a built-in support rod holder with locking knob that accepts a 1,3 cm (¹/₂") Ø support rod (sold separately)