VWR® Talon®, Support stand, U-shaped base
Supplier: VWR Collection
12985-078
VWRU12985-078EA 102 SGD
VWRU12985-078
VWR® Talon®, Support stand, U-shaped base
Retort Stands
Cast iron with black enamel finish.
  • Support stand base for use with VWR® Talon® Lab Frames
  • Three built-in support rod holders for additional support or to customise your stand
  • Locking knob accepts a 13 mm Ø support rod (sold separately)
Order Now
SPECIFICATIONS
Environmentally Preferable