VWR®, Beakers, Low Form, with Handle
Supplier: VWR Collection
13916-820 13916-795 13916-707 13916-762
VWRU13916-820EA 91.3 SGD
VWRU13916-820 VWRU13916-795 VWRU13916-707 VWRU13916-762
VWR®, Beakers, Low Form, with Handle
Beakers
PP, translucent.
  • Robust with non-drip spout and ridged thumb-grip on handle
  • Low form for better stability
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable