VWR®, Biohazard Bags
Supplier: VWR Collection
14220-006 14220-012 14220-010 14220-000 14220-002 14220-004
VWRU14220-006EA 246 SGD
VWRU14220-006 VWRU14220-012 VWRU14220-010 VWRU14220-000 VWRU14220-002 VWRU14220-004
VWR®, Biohazard Bags
Bags Waste Disposal Bags
These bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
  • PP, 38 µm thick
  • Perfect adaptability to standard laboratory disposal routines, including steam autoclaving
  • With biohazard warning and printed area
  • For use at 135 °C
Delivery information: Twist ties are included.
Order Now
SPECIFICATIONS
Environmentally Preferable