VWR®, Autoclavable Infectious Waste Bag, High Strength

Supplier: VWR Collection
14230-910
VWRU14230-910EA 248 SGD
VWRU14230-910
VWR®, Autoclavable Infectious Waste Bag, High Strength
Bags Waste Disposal Bags
These high strength bags, made from PP, are ideal for hazardous waste collection and decontamination in an autoclave. Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory. They are red and printed with a black biohazard symbol: 'Infectious Waste' and feature a temperature indicator patch.

  • Designed to handle dangerous materials
  • High-strength
  • Autoclavable to 135 °C (275 °F)

Certifications: Meets 480 g tear test (ASTM D1299-94A).

Delivery information: Supplied with 12,7 cm twist ties.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR