VWR®, Autoclavable Bags
Supplier: VWR Collection
14232-184
VWRU14232-184EA 269 SGD
VWRU14232-184 VWRI129-1575
VWR®, Autoclavable Bags
Bags Waste Disposal Bags
Sturdy, temperature resistant bags, ideal for sterilising animal cages.
  • PE, clear
  • Autoclavable up to 121 °C
  • Highly puncture resistant and impact strength
Order Now
SPECIFICATIONS
Environmentally Preferable