VWR®, Laboratory Jacks
Supplier: VWR Collection
14233-370 14233-366 14233-362 14233-368 14233-372
VWRU14233-370EA 2460 SGD
VWRU14233-370 VWRU14233-366 VWRU14233-362 VWRU14233-368 VWRU14233-372
VWR®, Laboratory Jacks
Laboratory Jacks
Stainless steel lab jacks are ideal for use in fume hoods or benchtops. These ultra-stable lifting platforms hold a variety of items such as glassware, hot plates, baths, and magnetic stirrers.
  • Constructed of 300-series stainless steel for exceptional strength and durability
  • Designed for use in extreme environments and high load applications
  • Durable construction allows jacks to be autoclaved or chemically cleaned
  • Equipped with oversized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments
  • Designed to hold the specified weight
More Product Information