VWR®, Staining Rack
Supplier: VWR Collection
15153-821
VWRU15153-821EA 188 SGD
VWRU15153-821
VWR®, Staining Rack
Racks Staining Racks
Stainless steel.
  • Triangular slide with a tapered top
  • Contact the slides only in a narrow zone
  • Simple and quickly adjustable
  • Fit sinks up to 534 mm inside dimensions
Staining rack is designed for use across laboratory sinks of various sizes.