VWR®, Microhematocrit Capillary Tubes
Supplier: VWR Collection
15401-560
VWRU15401-560EA 22.8 SGD
VWRU15401-560
VWR®, Microhematocrit Capillary Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Fire-polished, glass tubes.
  • Treated with heparin
  • Calibration line is present at 60 mm from tip
Order Now
SPECIFICATIONS
Environmentally Preferable