VWR®, Cover glasses for hemocytometer
Supplier: VWR Collection
16007-000
VWRU16007-000EA 74.5 SGD
VWRU16007-000
VWR®, Cover glasses for hemocytometer
Cover Glasses
Cover glasses with clipped corners.
  • Highly polished hard-ground glass