VWR®, Bottles, Wide Neck, Round, HDPE, with Screw Cap
Supplier: VWR Collection
16125-785 16125-967 16125-901
VWRU16125-785EA 45.9 SGD
VWRU16125-785 VWRU16125-967 VWRU16125-901
VWR®, Bottles, Wide Neck, Round, HDPE, with Screw Cap
Bottles Plastic Bottles
HDPE, natural.
These bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping. It provides an excellent moisture barrier and impact resistant.
Order Now
SPECIFICATIONS
Environmentally Preferable