VWR® Talon®, Burette clamp
Supplier: VWR Collection
17683-258
VWRU17683-258EA 131 SGD
VWRU17683-258
VWR® Talon®, Burette clamp
Clamps General Purpose Clamps
Zinc. This double burette clamp holds any size burette from micro to 100 ml capacity. Simply compress the scissor-like mechanism, insert burette, and gently release to grip.
  • Numbers and graduation on burette remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release