VWR® SuperSpin, Microcentrifuge Tubes, 1,5 ml
Supplier: VWR Collection
20170-026
VWRU20170-026EA 41.9 SGD
VWRU20170-026
VWR® SuperSpin, Microcentrifuge Tubes, 1,5 ml
Tubes Microcentrifuge Tubes
Ultra-clear PP, graduated, autoclavable and freezable with flat caps, easy to open. Amber coloured for light-sensitive substances.
  • Can withstand ultra-high centrifugation to RCF 40000×g
  • Fit all standard, high capacity rotors
  • Flat cap can be easily labelled and pierced, includes a special 90° hinge, making it easier to close while pipetting one-handed
  • Good for phenol/chloroform vortexing
  • Every lot is tested and guaranteed DNase- and RNase-free
  • Sustainable design based on virgin resins
Packaging: In resealable bags. Assorted colours pack includesMore Product Information