VWR®, Culture tube, disposable, with dual position cap
Supplier: VWR Collection
20170-579
VWRU20170-579EA 281 SGD
VWRU20170-579
VWR®, Culture tube, disposable, with dual position cap
Tubes Culture Tubes
PP, translucent.
  • Autoclavable at 122 °C
  • Freezable to –90 °C
  • Caps are not autoclavable
Tubes feature dual-position polyethylene caps, which permit sample aspiration.