VWR®, Racks for Microcentrifuge Tubes
Supplier: VWR Collection
20901-675
VWRU20901-675EA 65.1 SGD
VWRU20901-675
VWR®, Racks for Microcentrifuge Tubes
Racks Tube Racks
PP.
  • Easily transportable with one hand
  • Non-skid rubber feet
  • Autoclavable