VWR® SuperClear, Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
21008-240 82018-048 21008-178 21008-105
VWRU21008-240EA 0 SGD
VWRU21008-240 VWRU82018-048 VWRU21008-178 VWRU21008-105
VWR® SuperClear, Ultra-High Performance Centrifuge Tubes
Tubes Centrifuge Tubes
These popular, conical-bottom centrifuge tubes are designed for high speed centrifugation, and are made of non cytotoxic, medical grade resin that allows users to easily see sample volume and colour.
  • Prevents breakage and leakage during high speed centrifugation
  • Easy to read graduations for accurate measurements
  • Patented extra large, smear resistant writing panel
  • Versatile packaging options
  • Free from RNase, DNase, endotoxin and ATP
SuperClear tubes feature highly visible printed graduations for easy filling and sample preparation. The patented, extraMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M