VWR® Talon®, Swivel holder
Supplier: VWR Collection
21572-603
VWRU21572-603EA 72.4 SGD
VWRU21572-603
VWR® Talon®, Swivel holder
Clamps Bossheads
This Q-clamp post holder with center swivel capacity allows tilting of clamps at any angle in parallel planes. Made of nickel-plated zinc.
  • Allows tilting at any angle
  • Outside adjustment screw allows close proximity between objects being held