VWR® Talon®, Clamp holder
Supplier: VWR Collection
21572-658
VWRU21572-658EA 128 SGD
VWRU21572-658
VWR® Talon®, Clamp holder
Clamps Bossheads
Building on the upgraded Q-clamp design, this all-position clamp holder allows adjustment at any angle in any plane.
  • Holders are set at 90° to each other connected by a 90° connector for 360° rotation