VWR®, Alarm humidity/temperature monitor, remote, Traceable
Supplier: VWR Collection
21800-066
VWRU21800-066EA 350 SGD
VWRU21800-066
VWR®, Alarm humidity/temperature monitor, remote, Traceable
Thermometers Digital Thermometers
Triple display simultaneously shows high, low and current humidity/temperature readings. It is ideal for routine measurements, monitoring round-the-clock, all quality-control needs and critical experimental requirements. Used in hoods, classrooms, stockrooms, environmental chambers and chemical storage areas.
  • Hi/Lo alarms, min/max memories
  • High/low alarm feature may be set by user in 1% RH or 1° increments
Certifications: Compliance with ISO/IEC 17025:2005 (1750.01); accredited by A2LA indicating traceability to standards provided by NIST.