VWR®, Accessories for Bench Conductivity Meter
Supplier: VWR Collection
23226-504
VWRU23226-504EA 790 SGD
VWRU23226-504
VWR®, Accessories for Bench Conductivity Meter
Electrochemistry Meters Conductivity Meters
This glass probe is for use with the VWR® Traceable® conductivity bench meter (VWRU61161-362) or the VWR® bench/portable conductivity meter (VWRU23226-505).