VWR® Traceable®, Hand tally counter
Supplier: VWR Collection
23609-102
VWRU23609-102EA 64.9 SGD
VWRU23609-102
VWR® Traceable®, Hand tally counter
Cell Counters and Flow Cytometers Manual Cell Counters
Tally counter is designed to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples and tally biomedical events.
  • Rugged, heavy-duty chrome-finished case with hinged finger loop for grip
  • Audible click that signals the count
  • Four figures, easy-to-use
Delivery information: Supplied with a loop.
Order Now
SPECIFICATIONS
Environmentally Preferable