VWR® Traceable® Optik-Wipes™, Cleaning wipes for safety glasses
Supplier: VWR Collection
23609-168
VWRU23609-168EA 62.3 SGD
VWRU23609-168
VWR® Traceable® Optik-Wipes™, Cleaning wipes for safety glasses
Wipes Disposable Wipes and Towels Lens Papers
This strong, non woven cloth constructed of cellulose and polyester will not scratch surfaces or fall apart. It is pre-moistened with a unique formulation which easily removes oil, grease and lint.
  • Soft and lint-free
  • Clean smudges, dirt, dust, and the static that attracts them
  • Wipe material is 90%
  • Individually dispensed wipe is handy and ready for instant use
  • Each canister is provided in shrink-wrapped for use in clean rooms
  • Reclosable container assures a clean wipe every time and eliminates contamination from dust or dirt
Designed specifically forMore Product Information