VWR®, Spray Bottles, Adjustable
Supplier: VWR Collection
23609-182 16650-049
VWRU23609-182EA 78.4 SGD
VWRU23609-182 VWRU16650-049
VWR®, Spray Bottles, Adjustable
Bottles Spray Bottles
HDPE with PP spray cap.
  • Large capacity minimises refilling
  • Chemical-resistant and virtually unbreakable
  • Gas or chemically sterilised and dishwasher safe
  • Spray pattern can be controlled by a contoured spray trigger with 'instant off'
These wash bottles with adjustable-spray stream keep the contents completely free of contaminants. Virtually closed system eliminates drips and handles hot, cold, aqueous and solvent-based solutions.