VWR®, Staining Jars according to Hellendahl
Supplier: VWR Collection
25460-907
VWRU25460-907EA 49.7 SGD
VWRU25460-907
VWR®, Staining Jars according to Hellendahl
Jars Staining Jars
PMP, with cover.
Holds 76×26 mm slides vertically. Eight grooves accommodate 16 slides back to back. Xylene can cause etching after 2 days at room temperature.