VWR® Syringe Filters
Supplier: VWR Collection
28145-495
VWRU28145-495EA 0 SGD
VWRU28145-495
VWR® Syringe Filters
Filters Syringe Filters
PP, nylon and PTFE membranes are ideal for sample preparation and small volume chemical filtration. Syringe filters with CA or PES membranes are ideal for sterile filtration of buffers, tissue culture media or additives and other biologicals. The PES membrane offers high flow rates. Non sterile syringe filters are excellent for HPLC applications. Each filter has a female Luer lock inlet and a male slip Luer outlet.
  • Manufacturing process certified to ISO 9001
  • Biosafe according to Class VI plastics tests
  • Not intended for use with aqueous solutions