VWR®, Bibulous Paper
Supplier: VWR Collection
28511-007
VWRU28511-007EA 85 SGD
VWRU28511-007
VWR®, Bibulous Paper
Blotting Papers
High quality, highly absorbent paper, lint free, suitable for drying slides and small glass components.