VWR®, Ring Weights for Flasks
Supplier: VWR Collection
29700-026 29700-048 29700-004 29700-060
VWRU29700-026EA 78.4 SGD
VWRU29700-026 VWRU29700-048 VWRU29700-004 VWRU29700-060
VWR®, Ring Weights for Flasks
Weights Flask Weights
Vikem® vinyl coated lead rings prevent flasks floating during water bath incubations and help prevent spillages or breakages.
  • Bright red vinyl covering protects flask from scratches
  • Stabilise flasks in water baths or on the benchtop