VWR® Durac™, Accurate hydrometer for specific gravity
Supplier: VWR Collection
34627-413
VWRU34627-413EA 64.2 SGD
VWRU34627-413
VWR® Durac™, Accurate hydrometer for specific gravity
Hydrometers
These hydrometers are designed for determining the specific gravity of liquids in industries such as food and beverage, chemical, pharmacology, transportation and refrigeration.
  • Individually serialised
  • Features a lead free metal ballast
  • Temperature of standardisation of 15,6 °C
Certifications: Traceable to NIST and DAkkS. Calibration conforms with A2LA accreditions.
Delivery information: Supplied with a multi-language (English, French, German, Italian, Portuguese, Spanish) statement of accuracy.